Assay Details
| Gene Symbol: plcd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099423.1 | NP_001092893.1 | 8 - 9 | 1725 | 80 | |
| GenBank mRNA | BC139849.1 | - | 8 - 9 | 1725 | 80 | |
More Information
Set Membership: |
|

plcd3a
Genomic Map