Assay Details
Gene Symbol: atf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103192.2 | NP_001096662.1 | 2 - 3 | 554 | 84 | |
GenBank mRNA | BC151935.1 | - | 2 - 3 | 558 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
