Assay Details
Gene Symbol: kiss1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105679.2 | NP_001099149.2 | 1 - 2 | 224 | 80 | |
GenBank mRNA | AB623211.1 | - | 1 - 2 | 224 | 80 | |
EU047917.1 | - | 1 - 2 | 224 | 80 | ||
HQ222888.1 | - | 1 - 2 | 224 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
