Assay Details
| Gene Symbol: gc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109695.1 | NP_001103165.1 | 11 - 12 | 2428 | 96 | |
| XM_017356806.2 | XP_017212295.1 | 11 - 12 | 2428 | 96 | ||
| XM_021477384.1 | XP_021333059.1 | 12 - 13 | 2723 | 96 | ||
More Information
Set Membership: |
|

gc2
Genomic Map