Assay Details
| Gene Symbol: gc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109695.1 | NP_001103165.1 | 1 - 2 | 744 | 67 | |
| XM_017356806.2 | XP_017212295.1 | 1 - 2 | 744 | 67 | ||
| XM_021477384.1 | XP_021333059.1 | 2 - 3 | 1039 | 67 | ||
More Information
Set Membership: |
|

gc2
Genomic Map