Assay Details
| Gene Symbol: kcnk13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110117.2 | NP_001103587.1 | 1 - 2 | 533 | 101 | |
| GenBank mRNA | BC153996.1 | - | 1 - 2 | 535 | 101 | |
More Information
Set Membership: |
|

kcnk13a
Genomic Map