Assay Details
Gene Symbol: otx2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131215.3 | NP_571290.1 | 3 - 4 | 515 | 73 | |
GenBank mRNA | BC076432.1 | - | 3 - 4 | 519 | 73 | |
D26174.1 | - | 3 - 4 | 483 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
