Assay Details
| Gene Symbol: raraa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131406.2 | NP_571481.2 | 5 - 6 | 1405 | 66 | |
| XM_005156052.4 | XP_005156109.1 | 7 - 8 | 1389 | 66 | ||
| GenBank mRNA | BC092692.1 | - | 5 - 6 | 1405 | 66 | |
| KF148618.1 | - | 5 - 6 | 1023 | 66 | ||
| L03398.1 | - | 5 - 6 | 887 | 66 | ||
| S74155.1 | - | 5 - 6 | 976 | 66 | ||
More Information
Set Membership: |
|

raraa
Genomic Map