Assay Details
Gene Symbol: jag2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131862.1 | NP_571937.1 | 25 - 26 | 3396 | 71 | |
XM_005160485.4 | XP_005160542.1 | 24 - 25 | 3303 | 71 | ||
GenBank mRNA | AF090432.1 | - | 25 - 26 | 3379 | 71 | |
AF229449.1 | - | 25 - 26 | 3396 | 71 | ||
AF229450.1 | - | 24 - 25 | 3282 | 71 | ||
BC163889.1 | - | 25 - 26 | 3383 | 71 | ||
CU458993.1 | - | 25 - 26 | 3379 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
