Assay Details
| Gene Symbol: ddx55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173229.2 | NP_775336.2 | 8 - 9 | 908 | 69 | |
| GenBank mRNA | AF506203.1 | - | 8 - 9 | 873 | 69 | |
| BC059534.1 | - | 8 - 9 | 834 | 69 | ||
| EH441837.1 | - | 8 - 9 | 921 | 69 | ||
More Information
Set Membership: |
|

ddx55
Genomic Map