Assay Details
| Gene Symbol: c19h1orf109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173269.1 | NP_775376.1 | 2 - 3 | 546 | 50 | |
| GenBank mRNA | AF506221.1 | - | 2 - 3 | 546 | 50 | |
| BC094993.1 | - | 2 - 3 | 618 | 50 | ||
| BC164043.1 | - | 2 - 3 | 485 | 50 | ||
More Information
Set Membership: |
|

c19h1orf109
Genomic Map