Assay Details
| Gene Symbol: tph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178306.3 | NP_840091.2 | 1 - 2 | 153 | 104 | |
| GenBank mRNA | AF548566.1 | - | 1 - 2 | 165 | 104 | |
| BC059550.1 | - | 1 - 2 | 154 | 104 | ||
More Information
Set Membership: |
|

tph1a
Genomic Map