Assay Details
| Gene Symbol: tph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178306.3 | NP_840091.2 | 2 - 3 | 289 | 91 | |
| XM_017354058.2 | XP_017209547.1 | 2 - 3 | 265 | 91 | ||
| GenBank mRNA | AF548566.1 | - | 2 - 3 | 301 | 91 | |
| BC059550.1 | - | 2 - 3 | 290 | 91 | ||
More Information
Set Membership: |
|

tph1a
Genomic Map