Assay Details
| Gene Symbol: pdgfaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194426.3 | NP_919407.2 | 4 - 5 | 976 | 116 | |
| GenBank mRNA | AF200950.1 | - | 4 - 5 | 865 | 116 | |
| BC078289.1 | - | 4 - 5 | 116 | |||
More Information
Set Membership: |
|

pdgfaa
Genomic Map