Assay Details
| Gene Symbol: pdgfaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194426.3 | NP_919407.2 | 5 - 6 | 1096 | 89 | |
| GenBank mRNA | AF200950.1 | - | 5 - 6 | 985 | 89 | |
| BC078289.1 | - | 5 - 6 | 996 | 88 | ||
More Information
Set Membership: |
|

pdgfaa
Genomic Map