Assay Details
| Gene Symbol: hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198210.2 | NP_937853.1 | 4 - 5 | 495 | 74 | |
| GenBank mRNA | AF510108.1 | - | 4 - 5 | 475 | 74 | |
| BC063951.1 | - | 4 - 5 | 495 | 74 | ||
More Information
Set Membership: |
|

hsp90b1
Genomic Map