Assay Details
Gene Symbol: gspt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198806.1 | NP_942101.1 | 13 - 14 | 1835 | 73 | |
GenBank mRNA | BC053244.1 | - | 13 - 14 | 1835 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
