Assay Details
| Gene Symbol: gspt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198806.1 | NP_942101.1 | 7 - 8 | 1078 | 68 | |
| GenBank mRNA | BC053244.1 | - | 7 - 8 | 1078 | 68 | |
More Information
Set Membership: |
|

gspt1l
Genomic Map