Assay Details
Gene Symbol: nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199568.2 | NP_955862.2 | 4 - 5 | 713 | 71 | |
GenBank mRNA | BC046083.1 | - | 4 - 5 | 662 | 71 | |
EH457038.1 | - | 4 - 5 | 702 | 71 | ||
EH531450.1 | - | 2 - 3 | 471 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
