Assay Details
| Gene Symbol: mtmr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200394.1 | NP_956688.1 | 7 - 8 | 1007 | 80 | |
| XM_021476036.1 | XP_021331711.1 | 2 - 3 | 153 | 80 | ||
| GenBank mRNA | AY423041.1 | - | 7 - 8 | 934 | 80 | |
| BC053411.1 | - | 7 - 8 | 1007 | 80 | ||
More Information
Set Membership: |
|

mtmr8
Genomic Map