Assay Details
Gene Symbol: mtmr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200394.1 | NP_956688.1 | 10 - 11 | 1293 | 147 | |
XM_021476036.1 | XP_021331711.1 | 5 - 6 | 439 | 147 | ||
GenBank mRNA | AY423041.1 | - | 10 - 11 | 1220 | 147 | |
BC053411.1 | - | 10 - 11 | 1293 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
