Assay Details
| Gene Symbol: ptpn11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200454.1 | NP_956748.1 | 4 - 5 | 615 | 72 | |
| XM_005162157.4 | XP_005162214.1 | 4 - 5 | 653 | 72 | ||
| XM_009296835.3 | XP_009295110.1 | 4 - 5 | 807 | 72 | ||
| GenBank mRNA | BC055139.1 | - | 4 - 5 | 615 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009296835.3 transcripts.
Set Membership: |
|

ptpn11b
Genomic Map