Assay Details
| Gene Symbol: gmppab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200497.1 | NP_956791.1 | 7 - 8 | 989 | 73 | |
| GenBank mRNA | BC055506.1 | - | 7 - 8 | 989 | 73 | |
More Information
Set Membership: |
|

gmppab
Genomic Map