Assay Details
| Gene Symbol: ndufv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200747.1 | NP_957041.1 | 7 - 8 | 683 | 58 | |
| GenBank mRNA | BC059546.1 | - | 7 - 8 | 683 | 58 | |
More Information
Set Membership: |
|

ndufv2
Genomic Map