Assay Details
| Gene Symbol: kcnip3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200819.1 | NP_957113.1 | 6 - 7 | 639 | 89 | |
| XM_005169088.4 | XP_005169145.1 | 7 - 8 | 775 | 89 | ||
| XM_009304180.3 | XP_009302455.1 | 6 - 7 | 766 | 89 | ||
| XM_009304181.3 | XP_009302456.1 | 6 - 7 | 1629 | 89 | ||
| XM_009304183.3 | XP_009302458.1 | 5 - 6 | 1622 | 89 | ||
| XM_009304184.3 | XP_009302459.1 | 5 - 6 | 556 | 89 | ||
| XM_021478217.1 | XP_021333892.1 | 5 - 6 | 448 | 89 | ||
| GenBank mRNA | BC059703.1 | - | 6 - 7 | 639 | 89 | |
More Information
Set Membership: |
|

kcnip3a
Genomic Map