Assay Details
Gene Symbol: prkcbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200978.1 | NP_957272.1 | 16 - 17 | 2002 | 61 | |
GenBank mRNA | BC055154.1 | - | 16 - 17 | 2002 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
