Assay Details
| Gene Symbol: prkcbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200978.1 | NP_957272.1 | 7 - 8 | 967 | 93 | |
| XM_005164005.4 | XP_005164062.1 | 7 - 8 | 994 | 93 | ||
| GenBank mRNA | AY130432.1 | - | 5 - 6 | 619 | 93 | |
| BC055154.1 | - | 7 - 8 | 967 | 93 | ||
More Information
Set Membership: |
|

prkcbb
Genomic Map