Assay Details
Gene Symbol: prkcbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200978.1 | NP_957272.1 | 9 - 10 | 1203 | 105 | |
XM_005164005.4 | XP_005164062.1 | 9 - 10 | 1230 | 105 | ||
GenBank mRNA | AY130432.1 | - | 7 - 8 | 855 | 105 | |
BC055154.1 | - | 9 - 10 | 1203 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
