Assay Details
Gene Symbol: cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201084.2 | NP_957378.2 | 8 - 9 | 1252 | 60 | |
GenBank mRNA | BC044511.1 | - | 8 - 9 | 1252 | 60 | |
BC165281.1 | - | 8 - 9 | 1127 | 60 | ||
DY544594.1 | - | 1 - 2 | 27 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
