Assay Details
| Gene Symbol: seta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201475.1 | NP_958883.1 | 4 - 5 | 580 | 110 | |
| GenBank mRNA | BC046082.1 | - | 4 - 5 | 580 | 110 | |
More Information
Set Membership: |
|

seta
Genomic Map