Assay Details
| Gene Symbol: csrp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205567.1 | NP_991130.1 | 5 - 6 | 623 | 92 | |
| GenBank mRNA | BC065956.1 | - | 5 - 6 | 623 | 92 | |
More Information
Set Membership: |
|

csrp1a
Genomic Map