Assay Details
| Gene Symbol: cry-dash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205686.1 | NP_991249.1 | 11 - 12 | 1165 | 69 | |
| GenBank mRNA | AB120759.1 | - | 11 - 12 | 1165 | 69 | |
| BC098514.1 | - | 11 - 12 | 1235 | 69 | ||
More Information
Set Membership: |
|

cry-dash
Genomic Map