Assay Details
Gene Symbol: cry-dash | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205686.1 | NP_991249.1 | 1 - 2 | 66 | 68 | |
GenBank mRNA | AB120759.1 | - | 1 - 2 | 66 | 68 | |
BC098514.1 | - | 1 - 2 | 136 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
