Assay Details
| Gene Symbol: cry-dash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205686.1 | NP_991249.1 | 7 - 8 | 772 | 117 | |
| GenBank mRNA | AB120759.1 | - | 7 - 8 | 772 | 117 | |
| BC098514.1 | - | 7 - 8 | 842 | 117 | ||
More Information
Set Membership: |
|

cry-dash
Genomic Map