Assay Details
| Gene Symbol: cry-dash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205686.1 | NP_991249.1 | 9 - 10 | 943 | 101 | |
| GenBank mRNA | AB120759.1 | - | 9 - 10 | 943 | 101 | |
| BC098514.1 | - | 9 - 10 | 1013 | 101 | ||
More Information
Set Membership: |
|

cry-dash
Genomic Map