Assay Details
| Gene Symbol: yy1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212617.2 | NP_997782.1 | 3 - 4 | 849 | 69 | |
| GenBank mRNA | AF389345.1 | - | 1720 | 69 | ||
| BC071351.1 | - | 3 - 4 | 849 | 69 | ||
| HQ166834.1 | - | 3 - 4 | 733 | 69 | ||
More Information
Set Membership: |
|

yy1a
Genomic Map