Assay Details
| Gene Symbol: ptpn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212654.1 | NP_997819.1 | 3 - 4 | 385 | 94 | |
| GenBank mRNA | BC044373.1 | - | 3 - 4 | 385 | 94 | |
| BC066698.1 | - | 3 - 4 | 94 | |||
| BC164031.1 | - | 3 - 4 | 289 | 94 | ||
More Information
Set Membership: |
|

ptpn2b
Genomic Map