Assay Details
| Gene Symbol: ptpn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212654.1 | NP_997819.1 | 5 - 6 | 619 | 110 | |
| GenBank mRNA | AF097483.1 | - | 1 - 2 | 126 | 110 | |
| BC044373.1 | - | 5 - 6 | 619 | 110 | ||
| BC066698.1 | - | 5 - 6 | 612 | 110 | ||
| BC164031.1 | - | 5 - 6 | 523 | 110 | ||
More Information
Set Membership: |
|

ptpn2b
Genomic Map