Assay Details
| Gene Symbol: cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212685.1 | NP_997850.1 | 7 - 8 | 728 | 88 | |
| GenBank mRNA | BC066624.1 | - | 7 - 8 | 728 | 88 | |
More Information
Set Membership: |
|

cyb5r3
Genomic Map