Assay Details
Gene Symbol: ddx39ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212977.2 | NP_998142.1 | 2 - 3 | 335 | 57 | |
GenBank mRNA | BC044169.1 | - | 2 - 3 | 305 | 57 | |
BC067555.1 | - | 2 - 3 | 275 | 57 | ||
BC128804.1 | - | 2 - 3 | 280 | 57 | ||
BC165847.1 | - | 1 - 2 | 238 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
