Assay Details
Gene Symbol: ddx39ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212977.2 | NP_998142.1 | 4 - 5 | 750 | 67 | |
GenBank mRNA | BC044169.1 | - | 4 - 5 | 720 | 67 | |
BC067555.1 | - | 4 - 5 | 690 | 67 | ||
BC128804.1 | - | 4 - 5 | 695 | 67 | ||
BC165847.1 | - | 3 - 4 | 653 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
