Assay Details
| Gene Symbol: naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213334.1 | NP_998499.1 | 3 - 4 | 297 | 74 | |
| GenBank mRNA | BC053180.1 | - | 3 - 4 | 297 | 74 | |
| BC165054.1 | - | 3 - 4 | 213 | 74 | ||
More Information
Set Membership: |
|

naa10
Genomic Map