Assay Details
Gene Symbol: hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213426.1 | NP_998591.1 | 2 - 3 | 321 | 62 | |
GenBank mRNA | BC053413.1 | - | 2 - 3 | 321 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
