Assay Details
| Gene Symbol: ube2na | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213486.1 | NP_998651.1 | 2 - 3 | 388 | 84 | |
| GenBank mRNA | BC044461.1 | - | 2 - 3 | 388 | 84 | |
| BC164904.1 | - | 2 - 3 | 316 | 84 | ||
More Information
Set Membership: |
|

ube2na
Genomic Map