Assay Details
| Gene Symbol: aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213494.2 | NP_998659.2 | 3 - 4 | 870 | 55 | |
| GenBank mRNA | BC055234.1 | - | 3 - 4 | 467 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aebp2| Gene Symbol: aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213494.2 | NP_998659.2 | 3 - 4 | 870 | 55 | |
| GenBank mRNA | BC055234.1 | - | 3 - 4 | 467 | 55 | |
Set Membership: |
|