Assay Details
Gene Symbol: cant1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214743.1 | NP_999908.1 | 2 - 3 | 744 | 65 | |
XM_005171642.3 | XP_005171699.1 | 2 - 3 | 736 | 65 | ||
GenBank mRNA | BC068371.1 | - | 2 - 3 | 744 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
