Assay Details
| Gene Symbol: asic1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214789.1 | NP_999954.1 | 3 - 4 | 857 | 88 | |
| XM_017353815.2 | XP_017209304.1 | 3 - 4 | 1050 | 88 | ||
| XM_021470127.1 | XP_021325802.1 | 3 - 4 | 1054 | 88 | ||
| XM_021470128.1 | XP_021325803.1 | 3 - 4 | 1057 | 88 | ||
| XM_021470129.1 | XP_021325804.1 | 3 - 4 | 1611 | 88 | ||
| XM_021470130.1 | XP_021325805.1 | 3 - 4 | 1053 | 88 | ||
| GenBank mRNA | AJ609617.1 | - | 3 - 4 | 857 | 88 | |
More Information
Set Membership: |
|

asic1c
Genomic Map