Assay Details
| Gene Symbol: uqcrc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001589.1 | NP_001001589.1 | 11 - 12 | 1108 | 77 | |
| GenBank mRNA | BC054137.1 | - | 11 - 12 | 1086 | 77 | |
| BC071551.1 | - | 11 - 12 | 1108 | 77 | ||
| BC097011.1 | - | 11 - 12 | 1067 | 77 | ||
| BC152592.1 | - | 11 - 12 | 1086 | 77 | ||
More Information
Set Membership: |
|

uqcrc2b
Genomic Map