Assay Details
| Gene Symbol: uqcrc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001589.1 | NP_001001589.1 | 8 - 9 | 728 | 99 | |
| GenBank mRNA | BC071551.1 | - | 8 - 9 | 728 | 99 | |
More Information
Set Membership: |
|

uqcrc2b
Genomic Map