Assay Details
| Gene Symbol: uqcrc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001589.1 | NP_001001589.1 | 9 - 10 | 827 | 63 | |
| GenBank mRNA | BC054137.1 | - | 9 - 10 | 805 | 63 | |
| BC071551.1 | - | 9 - 10 | 827 | 63 | ||
| BC097011.1 | - | 9 - 10 | 786 | 63 | ||
| BC152592.1 | - | 9 - 10 | 805 | 63 | ||
More Information
Set Membership: |
|

uqcrc2b
Genomic Map